missing translation for 'onlineSavingsMsg'
Learn More

Dibenzo-15-crown 5-Ether 95.0+%, TCI America™

Catalog No. D28771G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D28771G Supplier TCI America Supplier No. D28771G
Only null left

Chemical Identifiers

CAS 14262-60-3
Molecular Formula C18H20O5
Molecular Weight (g/mol) 316.35
MDL Number MFCD00055949
InChI Key QSBFECWPKSRWNM-UHFFFAOYSA-N
Synonym dibenzo-15-crown-5, dibenzo-15-crown 5-ether, 6,7,9,10,17,18-hexahydrodibenzo b,h 1,4,7,10,13 pentaoxacyclopentadecine, diabenzo-15-crown-5, dibenzo-15-crown-5-ether, 23dibenzo15crown5ether, dibenzo-15-crown gc, 5,6,7,8,9,10,11,16,17,18,19-undecahydrodibenzo a,g 15 annulene, dibenzo b,h 1,4,7,10,13 pentaoxacyclopentadecin,6,7,9,10,17,18-hexahydro, 2,5,12,15,18-pentaoxatricyclo 17.4.0.0?, 1 1 tricosa-1 19 ,6,8,10,20,22-hexaene
PubChem CID 373245
IUPAC Name 2,5,12,15,18-pentaoxatricyclo[17.4.0.0⁶,¹¹]tricosa-1(23),6,8,10,19,21-hexaene
SMILES C1COC2=CC=CC=C2OCCOC2=CC=CC=C2OCCO1

Specifications

Melting Point 115°C
Quantity 1 g
Formula Weight 316.35
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzo-15-crown 5-Ether