accessibility menu, dialog, popup

UserName

OOOOO

CAS RN 14262-60-3

IUPAC Name: 2,5,12,15,18-pentaoxatricyclo[17.4.0.0⁶,¹¹]tricosa-1(23),6,8,10,19,21-hexaene
Synonyms: 23DIBENZO15CROWN5ether 2,3-dibenzo-15-crown-5-ether
Molecular Weight (g/mol): 316.35
Molecular Formula: C18H20O5
InChi Key: QSBFECWPKSRWNM-UHFFFAOYSA-N
SMILES: C1COC2=CC=CC=C2OCCOC2=CC=CC=C2OCCO1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results