missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzenesulfonimide 98.0+%, TCI America™
$294.52 - $857.46
Chemical Identifiers
| CAS | 2618-96-4 |
|---|---|
| Molecular Formula | C12H11NO4S2 |
| Molecular Weight (g/mol) | 297.343 |
| MDL Number | MFCD00025019 |
| InChI Key | OVQABVAKPIYHIG-UHFFFAOYSA-N |
| Synonym | dibenzenesulfonimide, benzenesulfonamide, n-phenylsulfonyl, dibenzenesulfonamide, bis phenylsulfonyl amine, n-phenylsulfonyl benzenesulfonamide, dibenzolsulfimid, dibenzolsulfimide, diphenylsulfonimide, benzenesulfonic acid imide, n-benzenesulfonyl benzenesulfonamide |
| PubChem CID | 75671 |
| IUPAC Name | N-(benzenesulfonyl)benzenesulfonamide |
| SMILES | C1=CC=C(C=C1)S(=O)(=O)NS(=O)(=O)C2=CC=CC=C2 |
Chemical Identifiers
| 2618-96-4 | |
| 297.343 | |
| OVQABVAKPIYHIG-UHFFFAOYSA-N | |
| 75671 | |
| C1=CC=C(C=C1)S(=O)(=O)NS(=O)(=O)C2=CC=CC=C2 |
| C12H11NO4S2 | |
| MFCD00025019 | |
| dibenzenesulfonimide, benzenesulfonamide, n-phenylsulfonyl, dibenzenesulfonamide, bis phenylsulfonyl amine, n-phenylsulfonyl benzenesulfonamide, dibenzolsulfimid, dibenzolsulfimide, diphenylsulfonimide, benzenesulfonic acid imide, n-benzenesulfonyl benzenesulfonamide | |
| N-(benzenesulfonyl)benzenesulfonamide |
Specifications
| 2618-96-4 | |
| White-Yellow | |
| MFCD00025019 | |
| dibenzenesulfonimide, benzenesulfonamide, n-phenylsulfonyl, dibenzenesulfonamide, bis phenylsulfonyl amine, n-phenylsulfonyl benzenesulfonamide, dibenzolsulfimid, dibenzolsulfimide, diphenylsulfonimide, benzenesulfonic acid imide, n-benzenesulfonyl benzenesulfonamide | |
| C1=CC=C(C=C1)S(=O)(=O)NS(=O)(=O)C2=CC=CC=C2 | |
| 297.343 | |
| 297.34 | |
| Crystalline Powder |
| 158°C | |
| C12H11NO4S2 | |
| 5 g | |
| OVQABVAKPIYHIG-UHFFFAOYSA-N | |
| N-(benzenesulfonyl)benzenesulfonamide | |
| 75671 | |
| ≥98.0% (HPLC) | |
| Dibenzenesulfonimide |
Safety and Handling
EINECSNumber : (3)-3133
TSCA : Yes