missing translation for 'onlineSavingsMsg'
Learn More

Dibasic Sodium Phosphate, Dodecahydrate, USP, 98-100.5%, Spectrum™ Chemical

Catalog No. p-7157844
Change view
Click to view available options
Quantity:
500 g
50 kg
Packaging:
Fiber Drum
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602151 500 g Poly Bottle
18602150 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18602151 Supplier Spectrum Chemical Mfg Cor Supplier No. S2056500GM
Only null left

S2056, 10039-32-4, Na2HPO4·12H2O

Spectrum™ Chemical Dibasic Sodium Phosphate, Dodecahydrate, USP is an electrolyte replenisher and with radio-protective activity. Spectrum Chemical USP products, including all USP Sodium Phosphate products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10039-32-4
Molecular Formula H25Na2O16P
Molecular Weight (g/mol) 358.14
InChI Key DGLRDKLJZLEJCY-UHFFFAOYSA-L
IUPAC Name disodium dodecahydrate hydrogen phosphate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 98 to 100.5%
Grade USP
Loss on Drying 55.0-64%
Packaging Poly Bottle