missing translation for 'onlineSavingsMsg'
Learn More

Dibasic Sodium Phosphate, Dodecahydrate, USP, 98-100.5%, Spectrum™ Chemical

Catalog No. 18602150
Change view
Click to view available options
Quantity:
500 g
50 kg
Packaging:
Fiber Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602150 50 kg Fiber Drum
18602151 500 g Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18602150 Supplier Spectrum Chemical Mfg Cor Supplier No. S205650KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S2056, 10039-32-4, Na2HPO4·12H2O

Spectrum™ Chemical Dibasic Sodium Phosphate, Dodecahydrate, USP is an electrolyte replenisher and with radio-protective activity. Spectrum Chemical USP products, including all USP Sodium Phosphate products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10039-32-4
Molecular Formula H25Na2O16P
Molecular Weight (g/mol) 358.14
InChI Key DGLRDKLJZLEJCY-UHFFFAOYSA-L
IUPAC Name disodium dodecahydrate hydrogen phosphate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

CAS Max % 100%
Physical Form Crystals or Needles
Quantity 50 kg
Percent Purity 98 to 100.5%
Grade USP
Loss on Drying 55.0-64%
Packaging Fiber Drum
Chemical Name or Material Dibasic Sodium Phosphate, Dodecahydrate