missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibasic Calcium Phosphate, Dihydrate, Powder, USP, 98-105%, Spectrum™ Chemical
CA160, 7789-77-7, CaHPO4·2H2O
$532.40 - $6,570.80
Chemical Identifiers
| CAS | 7789-77-7 |
|---|---|
| Molecular Formula | CaH5O6P |
| Molecular Weight (g/mol) | 172.09 |
| InChI Key | XAAHAAMILDNBPS-UHFFFAOYSA-L |
| IUPAC Name | calcium dihydrate hydrogen phosphate |
| SMILES | O.O.[Ca++].OP([O-])([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18602140
|
Spectrum Chemical Mfg Cor
CA160500GM |
500 g | Poly Bottle |
N/A
|
|
||||
|
18602141
|
Spectrum Chemical Mfg Cor
CA16025KG |
2.5 kg | Poly Pail |
N/A
|
|
||||
|
18602142
|
Spectrum Chemical Mfg Cor
CA16012KG |
12 kg | Poly Pail |
N/A
|
|
||||
|
18602143
|
Spectrum Chemical Mfg Cor
CA16050KGBL |
50 kg | Fiber Drum |
N/A
|
|
||||
Description
Spectrum™ Chemical Dibasic Calcium Phosphate, Dihydrate, Powder, USP is used as a dietary supplement. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 7789-77-7 | |
| 172.09 | |
| calcium dihydrate hydrogen phosphate |
| CaH5O6P | |
| XAAHAAMILDNBPS-UHFFFAOYSA-L | |
| O.O.[Ca++].OP([O-])([O-])=O |
Specifications
| 7789-77-7 | |
| CaH5O6P | |
| XAAHAAMILDNBPS-UHFFFAOYSA-L | |
| calcium dihydrate hydrogen phosphate | |
| 98 to 105% | |
| Poly Bottle |
| 100% | |
| 500 g | |
| O.O.[Ca++].OP([O-])([O-])=O | |
| 172.09 | |
| USP |