missing translation for 'onlineSavingsMsg'
Learn More

Dibasic Calcium Phosphate, Dihydrate, Powder, USP, 98-105%, Spectrum™ Chemical

Catalog No. 18602140
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602140 500 g Poly Bottle
18602141 2.5 kg Poly Pail
18602142 12 kg Poly Pail
18602143 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602140 Supplier Spectrum Chemical Mfg Cor Supplier No. CA160500GM

CA160, 7789-77-7, CaHPO4·2H2O

Spectrum™ Chemical Dibasic Calcium Phosphate, Dihydrate, Powder, USP is used as a dietary supplement. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7789-77-7
Molecular Formula CaH5O6P
Molecular Weight (g/mol) 172.09
InChI Key XAAHAAMILDNBPS-UHFFFAOYSA-L
IUPAC Name calcium dihydrate hydrogen phosphate
SMILES O.O.[Ca++].OP([O-])([O-])=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 98 to 105%
Grade USP
Packaging Poly Bottle