missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Diazoxide, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 100 mg |
| Description | White to off-white |
| Content And Storage | 2°C to 8°C |
| Molecular Formula | C8H7ClN2O2S |
| Formula Weight | 230.67 |
| Synonym | 7-Chloro-3-methyl-2H-1,2,4-benzothiadiazine 1,1-dioxide |
| Melting Point | 330°C to 331°C |
| Solubility | Soluble in 0.1M NaOH. Insoluble in water or in methanol |
| Product Type | Diazoxide, 98% |
| Form | Powder |
| Show More |
Safety and Handling
| EINECSNumber | 206-668-1 |
| Recommended Storage | Keep cold |
RUO – Research Use Only