missing translation for 'onlineSavingsMsg'
Learn More

Diaveridine, 99.0% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111014512
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014512 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014512 Supplier Merck Emd Millipore Supplier No. D9516250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 5355-16-8
Physical Form Neat
Linear Formula C13H16N4O2
MDL Number MFCD00057349
Synonym 5-(3,4-Dimethoxybenzyl)-2,4-pyrimidinediamine
Type Analytical Standard
Quantity 250 mg
Formula Weight 260.29
Percent Purity ≥99% (TLC)
Grade Analytical Standard