missing translation for 'onlineSavingsMsg'
Learn More

Diatrizoic acid

Catalog No. AAJ6344414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6344414 25 g
AAJ6344422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6344414 Supplier Thermo Scientific Chemicals Supplier No. J6344414

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A high-osmolality radiocontrast agent | CAS: 117-96-4 | C11H9I3N2O4 | 613.92 g/mol

Diatrizoic acid is a gradient reagent used for blood cell separation and diagnostic aid (radiopaque medium). Diatrizoic acid may be used as an alternative to barium sulfate for medical imaging of the gastrointestinal tract, such as upper gastrointestinal series and small bowel series.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diatrizoic acid is a gradient reagent used for blood cell separation and diagnostic aid (radiopaque medium). Diatrizoic acid may be used as an alternative to barium sulfate for medical imaging of the gastrointestinal tract, such as upper gastrointestinal series and small bowel series.

Solubility
Soluble in Methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C. Keep away from oxidizing agents.

Chemical Identifiers

CAS 117-96-4
Molecular Formula C11H9I3N2O4
Molecular Weight (g/mol) 613.92
MDL Number MFCD00069960
InChI Key YVPYQUNUQOZFHG-UHFFFAOYSA-N
Synonym diatrizoic acid, amidotrizoic acid, diatrizoate, amidotrizoate, urogranoic acid, urografin acid, odiston, urotrast, diat, diatriazoate
PubChem CID 2140
ChEBI CHEBI:53691
IUPAC Name 3,5-diacetamido-2,4,6-triiodobenzoic acid
SMILES CC(=O)NC1=C(I)C(C(O)=O)=C(I)C(NC(C)=O)=C1I

Specifications

Color White
Quantity 25 g
Merck Index 14,2992
Solubility Information Soluble in Methanol.
Formula Weight 613.9
Physical Form Powder
Chemical Name or Material Diatrizoic acid
EINECSNumber 204-223-6
RTECSNumber DG5950000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only