missing translation for 'onlineSavingsMsg'
Learn More

Diamylammonium Acetate (ca. 0.5mol/L in Water) [Ion-Pair Reagent for LC-MS], TCI America™

Catalog No. A570410ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A570410ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A570410ML Supplier TCI America Supplier No. A570410ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 211676-91-4
Molecular Formula C12H27NO2
Molecular Weight (g/mol) 217.35
MDL Number MFCD03093618
InChI Key IPBJPAHGLXFUDS-UHFFFAOYSA-N
Synonym IPC-DAAA
PubChem CID 16218596
IUPAC Name acetic acid; dipentylamine
SMILES CC(O)=O.CCCCCNCCCCC

Specifications

Color Colorless
Quantity 10 mL
Formula Weight 217.35
Physical Form Liquid
Chemical Name or Material Diamylammonium Acetate (ca. 0.5mol/L in Water) [Ion-Pair Reagent for LC-MS]