accessibility menu, dialog, popup

UserName

CAS RN 211676-91-4

H3CNHCH3CH3OHO

CAS RN 211676-91-4

IUPAC Name: acetic acid; dipentylamine
Synonyms: acetic acid; dipentylamine
Molecular Weight (g/mol): 217.35
Molecular Formula: C12H27NO2
InChi Key: IPBJPAHGLXFUDS-UHFFFAOYSA-N
SMILES: CC(O)=O.CCCCCNCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results