missing translation for 'onlineSavingsMsg'
Learn More

Diamyl Phthalate 98.0+%, TCI America™

Catalog No. P029125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P029125G 25 g
P0291100G 100 g
P0291500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P029125G Supplier TCI America Supplier No. P029125G

Chemical Identifiers

CAS 131-18-0
Molecular Formula C18H26O4
Molecular Weight (g/mol) 306.402
MDL Number MFCD00041934
InChI Key IPKKHRVROFYTEK-UHFFFAOYSA-N
Synonym dipentyl phthalate, di-n-pentyl phthalate, diamyl phthalate, amyl phthalate, amoil, di-n-amyl phthalate, di-n-pentylphthalate, phthalic acid, dipentyl ester, phthalic acid diamyl ester, 1,2-benzenedicarboxylic acid, dipentyl ester
PubChem CID 8561
ChEBI CHEBI:34680
IUPAC Name dipentyl benzene-1,2-dicarboxylate
SMILES CCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCC

Specifications

Color Yellow
Boiling Point 342°C
Quantity 25 g
UN Number 3082
Formula Weight 306.40
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diamyl Phthalate
RTECSNumber TI1930000
TSCA Yes