missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Diamond PosiStop Pipettors Starter Pack - Adjustable Volume

Catalog No. 23763076
Change view
Click to view available options
Compatible Tips:
Rainin LTS Filter Tips
Rainin LTS Tips
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Compatible Tips
23763076 Rainin LTS Filter Tips
23763075 Rainin LTS Tips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23763076 Supplier Globe Scientific Supplier No. 3361COMBOFT
Only null left

Discover the Diamond PosiStop LTS-compatible pipettors - engineered for precision, comfort, and durability. With a low-force plunger, shock-absorbing tip ejector, and robust PVDF construction, they deliver consistent performance and ergonomic ease. Backed by a 3-year warranty.

  • Fully Autoclavable and UV Resistant: Ensuring durability and adaptability in various laboratory environments.
  • 3-Year Warranty: Providing extended assurance of quality and performance.
  • Color-Coded Plungers: For quick and easy volume identification, streamlining your laboratory processes.
  • Low Plunger Force: Offering a virtually fatigue-free pipetting experience.
  • Magnetically Assisted Piston: Guarantees extremely high accuracy and consistency.
  • Ergonomically Designed and Lightweight: Enhances comfort during prolonged use.
  • Precise Volume Adjustment: Allows for exact measurements and control.
  • Secure Locking Mechanism: Prevents accidental volume changes, ensuring reliable results.

Diamond PosiStop Pipettor Starter Pack:

  • Adjustable Volume Pipettors: 2 to 20 μL, 20 to 200 μL, 100 to 1000 μL
  • Racked Sterile Tips or Filter TIps: 20 μL, 200 μL, 1000 μL
  • 6-Place Carousel Stand
  • 10 mL, 25 mL, 50 mL Reagent Reservoirs

Specifications

Compatible Tips Rainin LTS Filter Tips
Autoclavable Autoclavable
Color Assorted
Disposable Reusable
For Use With Universal Pipette Tip
Grip Type Ergonomic
Material ABS Stand
Number of Channels 1
Sterility Non-sterile
Type Pipette Package, Adjustable
Volume (Metric) 2 to 1000 μL
Includes Adjustable Volume Pipettors, Racked Sterile Filter Tips, 6-Place Carousel Stand, Reagent Reservoirs
Show More Show Less