missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Globe Scientific Diamond PosiStop Pipettors Starter Pack - Adjustable Volume
Description
- Fully Autoclavable and UV Resistant: Ensuring durability and adaptability in various laboratory environments.
- 3-Year Warranty: Providing extended assurance of quality and performance.
- Color-Coded Plungers: For quick and easy volume identification, streamlining your laboratory processes.
- Low Plunger Force: Offering a virtually fatigue-free pipetting experience.
- Magnetically Assisted Piston: Guarantees extremely high accuracy and consistency.
- Ergonomically Designed and Lightweight: Enhances comfort during prolonged use.
- Precise Volume Adjustment: Allows for exact measurements and control.
- Secure Locking Mechanism: Prevents accidental volume changes, ensuring reliable results.
Diamond PosiStop Pipettor Starter Pack:
- Adjustable Volume Pipettors: 2 to 20 μL, 20 to 200 μL, 100 to 1000 μL
- Racked Sterile Tips or Filter TIps: 20 μL, 200 μL, 1000 μL
- 6-Place Carousel Stand
- 10 mL, 25 mL, 50 mL Reagent Reservoirs
Specifications
Specifications
| Compatible Tips | Rainin LTS Tips |
| Autoclavable | Autoclavable |
| Color | Assorted |
| Disposable | Reusable |
| For Use With | Universal Pipette Tip |
| Grip Type | Ergonomic |
| Material | ABS Stand |
| Number of Channels | 1 |
| Sterility | Non-sterile |
| Type | Pipette Package, Adjustable |
| Show More |