missing translation for 'onlineSavingsMsg'
Learn More

Diamantane 98.0+%, TCI America™

Catalog No. D28765G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28765G 5 g
D28761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28765G Supplier TCI America Supplier No. D28765G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2292-79-7
Molecular Formula C14H20
Molecular Weight (g/mol) 188.314
MDL Number MFCD00171301
InChI Key ZICQBHNGXDOVJF-UHFFFAOYSA-N
Synonym Congressane, Pentacyclo[7.3.1.1(4,12).0(2,7).0(6,11)]tetradecane
PubChem CID 123154
ChEBI CHEBI:38223
SMILES C1C2CC3C4C1C5CC(C4)CC3C5C2

Specifications

Melting Point 245°C
Color White-Yellow
Quantity 5 g
Formula Weight 188.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diamantane