accessibility menu, dialog, popup

UserName

CAS RN 2292-79-7

IUPAC Name: pentacyclo[7.3.1.1⁴,¹².0²,⁷.0⁶,¹¹]tetradecane
Synonyms: diamantane congressane diadamantane
Molecular Weight (g/mol): 188.31
Molecular Formula: C14H20
InChi Key: ZICQBHNGXDOVJF-UHFFFAOYSA-N
SMILES: C1C2CC3C4CC5CC(C14)C(C2)C3C5
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results