missing translation for 'onlineSavingsMsg'
Learn More

Diallyldiphenylsilane 95.0+%, TCI America™

Catalog No. D37305G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D37305G Supplier TCI America Supplier No. D37305G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10519-88-7
Molecular Formula C18H20Si
Molecular Weight (g/mol) 264.443
MDL Number MFCD00026068
InChI Key ZODWTWYKYYGSFS-UHFFFAOYSA-N
Synonym diallyldiphenylsilane, diphenyldiallylsilane, silane, diphenyldi-2-propenyl, silane, diallyldiphenyl, benzene, 1,1'-di-2-propen-1-ylsilylene bis, diphenylbis prop-2-en-1-yl silane, acmc-1bwl8, diphenyl-bis prop-2-enyl silane, benzene,1,1'-di-2-propen-1-ylsilylene bis
PubChem CID 82695
IUPAC Name diphenyl-bis(prop-2-enyl)silane
SMILES C=CC[Si](CC=C)(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Color Colorless
Boiling Point 141°C
Quantity 5 g
Formula Weight 264.44
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diallyldiphenylsilane