missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diallyldimethylsilane 97.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 135°C |
| Quantity | 25 mL |
| UN Number | 1993 |
| Formula Weight | 140.30 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Diallyldimethylsilane |
Safety and Handling
| TSCA | Yes |