accessibility menu, dialog, popup

UserName

CH3SiCH3CH2H2C

CAS RN 1113-12-8

IUPAC Name: dimethylbis(prop-2-en-1-yl)silane
Synonyms: dimethylbis(prop-2-en-1-yl)silane
Molecular Weight (g/mol): 140.3
Molecular Formula: C8H16Si
InChi Key: ZDSFBVVBFMKMRF-UHFFFAOYSA-N
SMILES: C[Si](C)(CC=C)CC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results