missing translation for 'onlineSavingsMsg'
Learn More

Diallyl sulfide, 98%

Catalog No. AAL1097114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1097114 25 g
AAL1097122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1097114 Supplier Thermo Scientific Chemicals Supplier No. L1097114

CAS: 592-88-1 | C6H10S | 114.21 g/mol

Diallyl sulfide is a modulator of drug metabolizing enzyme P450 system and inducer of the phase II detoxifying enzyme GST. Diallyl sulfide is also an inhibitor of CYP2E1 and believed to prevent chemically-induced carcinogenesis in many tissues. It is also used as a food additive. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diallyl sulfide is a modulator of drug metabolizing enzyme P450 system and inducer of the phase II detoxifying enzyme GST. Diallyl sulfide is also an inhibitor of CYP2E1 and believed to prevent chemically-induced carcinogenesis in many tissues. It is also used as a food additive. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in alcohol, chloroform, ether, and carbon tetrachloride. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 592-88-1
Molecular Formula C6H10S
Molecular Weight (g/mol) 114.21
MDL Number MFCD00008658
InChI Key UBJVUCKUDDKUJF-UHFFFAOYSA-N
Synonym diallyl sulfide, allyl sulfide, oil garlic, diallylsulfide, thioallyl ether, 1-propene, 3,3'-thiobis, allyl monosulfide, diallyl thioether, diallyl monosulfide, diallyl sulphide
PubChem CID 11617
ChEBI CHEBI:4489
SMILES C=CCSCC=C

Specifications

Melting Point -83°C
Density 0.889
Boiling Point 137°C to 138°C
Flash Point 23°C (73°F)
Odor Unpleasant
Refractive Index 1.4895
Linear Formula CH2=CHCH2SCH2CH=CH2
Quantity 25 g
UN Number UN1993
Beilstein 1736016
Merck Index 14,297
Solubility Information Soluble in alcohol,chloroform,ether,and carbon tetrachloride. Insoluble in water.
IUPAC Name 3-prop-2-enylsulfanylprop-1-ene
Formula Weight 114.21
Percent Purity 98%
Chemical Name or Material Diallyl sulfide
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 209-775-1
RTECSNumber BC4900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only