missing translation for 'onlineSavingsMsg'
Learn More

Diallyl Isophthalate 98.0+%, TCI America™

Catalog No. I045025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I045025G 25 g
I0450500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I045025G Supplier TCI America Supplier No. I045025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1087-21-4
Molecular Formula C14H14O4
Molecular Weight (g/mol) 246.262
MDL Number MFCD00048174
InChI Key OOORLLSLMPBSPT-UHFFFAOYSA-N
Synonym Isophthalic Acid Diallyl Ester
PubChem CID 66183
IUPAC Name bis(prop-2-enyl) benzene-1,3-dicarboxylate
SMILES C=CCOC(=O)C1=CC(=CC=C1)C(=O)OCC=C

Specifications

Color Yellow
Boiling Point 163°C
Quantity 25 g
Formula Weight 246.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Diallyl Isophthalate
EINECSNumber (3)-1317&(3)-1325&(3)-1329
RTECSNumber CZ4230000
TSCA Yes
Recommended Storage Refrigerator