missing translation for 'onlineSavingsMsg'
Learn More

Diallyl 1,4-Cyclohexanedicarboxylate (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. p-7127903
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D376825G 25 g
D3768250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D376825G Supplier TCI America Supplier No. D376825G

Chemical Identifiers

CAS 20306-22-3
Molecular Formula C14H20O4
Molecular Weight (g/mol) 252.31
MDL Number MFCD00050452
InChI Key ZXTPJLLIHIDBKQ-UHFFFAOYSA-N
Synonym 1,4-Cyclohexanedicarboxylic Acid Diallyl Ester
PubChem CID 11265237
IUPAC Name bis(prop-2-enyl) cyclohexane-1,4-dicarboxylate
SMILES C=CCOC(=O)C1CCC(CC1)C(=O)OCC=C

Specifications

Color Yellow
Quantity 25 g
Formula Weight 252.31
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diallyl 1,4-Cyclohexanedicarboxylate (cis- and trans- mixture)
EINECSNumber (3)-4376
TSCA No