missing translation for 'onlineSavingsMsg'
Learn More

Diacetyl 98.0+%, TCI America™

Catalog No. B068225ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B068225ML 25 mL
B0682100ML 100 mL
B0682500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B068225ML Supplier TCI America Supplier No. B068225ML

Chemical Identifiers

CAS 431-03-8
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00008756
InChI Key QSJXEFYPDANLFS-UHFFFAOYSA-N
Synonym 2,3-butanedione, diacetyl, biacetyl, dimethylglyoxal, 2,3-diketobutane, dimethyl glyoxal, butanedione, dimethyl diketone, 2,3-butadione, 2,3-dioxobutane
PubChem CID 650
ChEBI CHEBI:16583
IUPAC Name butane-2,3-dione
SMILES CC(=O)C(=O)C

Specifications

Melting Point -4°C
Color Yellow
Boiling Point 88°C
Quantity 25 mL
UN Number 2346
Formula Weight 86.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diacetyl
EINECSNumber (2)-0563
RTECSNumber EK2625000
TSCA Yes
Recommended Storage Refrigerator