missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Diacetyl-L-tartaric Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 120°C |
| Color | White |
| Quantity | 25 g |
| Formula Weight | 234.16 |
| Percent Purity | ≥98.0% (T) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (-)-Diacetyl-L-tartaric Acid |
Safety and Handling
| TSCA | No |