accessibility menu, dialog, popup

UserName

H3COOOCH3OOHOOHO

CAS RN 51591-38-9

IUPAC Name: 2,3-bis(acetyloxy)butanedioic acid
Synonyms: 2,3-bis(acetyloxy)butanedioic acid
Molecular Weight (g/mol): 234.16
Molecular Formula: C8H10O8
InChi Key: DNISEZBAYYIQFB-UHFFFAOYNA-N
SMILES: CC(=O)OC(C(OC(C)=O)C(O)=O)C(O)=O
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results