missing translation for 'onlineSavingsMsg'
Learn More

(-)-Diacetyl-D-tartaric Anhydride 98.0+%, TCI America™

Catalog No. D26455G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26455G 5 g
D264525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26455G Supplier TCI America Supplier No. D26455G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 70728-23-3
Molecular Formula C8H8O7
Molecular Weight (g/mol) 216.145
MDL Number MFCD02093461
InChI Key XAKITKDHDMPGPW-WDSKDSINSA-N
PubChem CID 12731855
IUPAC Name [(3S,4S)-4-acetyloxy-2,5-dioxooxolan-3-yl] acetate
SMILES CC(=O)OC1C(C(=O)OC1=O)OC(=O)C

Specifications

Melting Point 134°C
Color White
Quantity 5 g
Formula Weight 216.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Diacetyl-D-tartaric Anhydride
TSCA No
Recommended Storage Freezer