accessibility menu, dialog, popup

UserName

CAS RN 70728-23-3

CH3OOOOOOOH3C

CAS RN 70728-23-3

IUPAC Name: 4-(acetyloxy)-2,5-dioxooxolan-3-yl acetate
Synonyms: 4-(acetyloxy)-2,5-dioxooxolan-3-yl acetate
Molecular Weight (g/mol): 216.15
Molecular Formula: C8H8O7
InChi Key: XAKITKDHDMPGPW-UHFFFAOYNA-N
SMILES: CC(=O)OC1C(OC(C)=O)C(=O)OC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results