missing translation for 'onlineSavingsMsg'
Learn More

Diacetone acrylamide, 99%

Catalog No. AAA1594018
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1594018 50 g
AAA1594030 250 g
AAA159400B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1594018 Supplier Thermo Scientific Chemicals Supplier No. A1594018

CAS: 2873-97-4 | C9H15NO2 | 169.22 g/mol

Diacetone acrylamide is used in epoxy resin, acryl emulsion, water based paints, waste water treatment, photosensitive resins, waterborne coatings and gel electrophoresis. It is also used in the manufacture of permanent press fabrics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diacetone acrylamide is used in epoxy resin, acryl emulsion, water based paints, waste water treatment, photosensitive resins, waterborne coatings and gel electrophoresis. It is also used in the manufacture of permanent press fabrics.

Solubility
Soluble in water, methanol, benzene and tetrahydrofuran.

Notes
Store in cool place. Hygroscopic. Incompatible with strong bases and strong oxidizing agents.

Chemical Identifiers

CAS 2873-97-4
Molecular Formula C9H15NO2
Molecular Weight (g/mol) 169.22
MDL Number MFCD00008788
InChI Key OMNKZBIFPJNNIO-UHFFFAOYSA-N
Synonym diacetone acrylamide, diacetoneacrylamide, n-1,1-dimethyl-3-oxobutyl acrylamide, 2-propenamide, n-1,1-dimethyl-3-oxobutyl, n-2-methyl-4-oxopentan-2-yl acrylamide, n-2-2-methyl-4-oxopentyl acrylamide, acrylamide, n-1,1-dimethyl-3-oxobutyl, n-1,1-dimethyl-3-oxobutyl-2-propenamide, acrylamide, n,n-diacetonyl, ccris 5898
PubChem CID 17888
IUPAC Name N-(2-methyl-4-oxopentan-2-yl)prop-2-enamide
SMILES CC(=O)CC(C)(C)NC(=O)C=C

Specifications

Melting Point 54°C to 56°C
Flash Point >110°C (230°F)
Quantity 50 g
Beilstein 1928444
Sensitivity Hygroscopic
Merck Index 14,2963
Solubility Information Soluble in water,methanol,benzene and tetrahydrofuran.
Formula Weight 169.23
Percent Purity 99%
Chemical Name or Material Diacetone acrylamide
Hazard Category H302
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 220-713-2
RTECSNumber AS3475000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only