missing translation for 'onlineSavingsMsg'
Learn More

Diacerein 98.0+%, TCI America™

$188.27 - $486.52

Chemical Identifiers

CAS 13739-02-1
Molecular Formula C19H12O8
Molecular Weight (g/mol) 368.30
MDL Number MFCD00468030
InChI Key TYNLGDBUJLVSMA-UHFFFAOYSA-N
Synonym diacerein, diacetylrhein, diacerhein, artrodar, fisiodar, 1,8-diacetoxy-3-carboxyanthraquinone, 4,5-diacetylrhein, diacerin, diacerein inn, diacereine french
PubChem CID 26248
IUPAC Name 4,5-bis(acetyloxy)-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid
SMILES CC(=O)OC1=CC=CC2=C1C(=O)C1=C(OC(C)=O)C=C(C=C1C2=O)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D40611G
View Documents
TCI America
D40611G
1 g
Each for $188.27
Only null left
 
D40615G
View Documents
TCI America
D40615G
5 g
Each for $486.52
Only null left
 
Description

Description

Specifications

Chemical Identifiers

13739-02-1
368.30
TYNLGDBUJLVSMA-UHFFFAOYSA-N
26248
CC(=O)OC1=CC=CC2=C1C(=O)C1=C(OC(C)=O)C=C(C=C1C2=O)C(O)=O
C19H12O8
MFCD00468030
diacerein, diacetylrhein, diacerhein, artrodar, fisiodar, 1,8-diacetoxy-3-carboxyanthraquinone, 4,5-diacetylrhein, diacerin, diacerein inn, diacereine french
4,5-bis(acetyloxy)-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid

Specifications

13739-02-1
Yellow
MFCD00468030
diacerein, diacetylrhein, diacerhein, artrodar, fisiodar, 1,8-diacetoxy-3-carboxyanthraquinone, 4,5-diacetylrhein, diacerin, diacerein inn, diacereine french
CC(=O)OC1=CC=CC2=C1C(=O)C1=C(OC(C)=O)C=C(C=C1C2=O)C(O)=O
368.30
368.30
Crystalline Powder
243°C
C19H12O8
1 g
TYNLGDBUJLVSMA-UHFFFAOYSA-N
4,5-bis(acetyloxy)-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid
26248
≥98.0% (HPLC)
Diacerein
Videos
Safety and Handling

Safety and Handling

RTECSNumber : CB8781000

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'