missing translation for 'onlineSavingsMsg'
Learn More

Diacerein 98.0+%, TCI America™

Catalog No. D40615G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40615G 5 g
D40611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40615G Supplier TCI America Supplier No. D40615G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13739-02-1
Molecular Formula C19H12O8
Molecular Weight (g/mol) 368.30
MDL Number MFCD00468030
InChI Key TYNLGDBUJLVSMA-UHFFFAOYSA-N
Synonym diacerein, diacetylrhein, diacerhein, artrodar, fisiodar, 1,8-diacetoxy-3-carboxyanthraquinone, 4,5-diacetylrhein, diacerin, diacerein inn, diacereine french
PubChem CID 26248
IUPAC Name 4,5-bis(acetyloxy)-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid
SMILES CC(=O)OC1=CC=CC2=C1C(=O)C1=C(OC(C)=O)C=C(C=C1C2=O)C(O)=O

Specifications

Melting Point 243°C
Color Yellow
Quantity 5 g
Formula Weight 368.30
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Diacerein
RTECSNumber CB8781000
TSCA No