missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-octyl Disulfide 75.0+%, TCI America™

Catalog No. D085825ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D085825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D085825ML Supplier TCI America Supplier No. D085825ML

Chemical Identifiers

CAS 29956-99-8
Molecular Formula C16H34S2
Molecular Weight (g/mol) 290.568
MDL Number MFCD00048262
InChI Key ZIMCZOLRXKPXLN-UHFFFAOYSA-N
Synonym tert-Octyl Disulfide, Bis(1,1,3,3-tetramethylbutyl) Disulfide
PubChem CID 121601
IUPAC Name 2,2,4-trimethyl-4-(2,4,4-trimethylpentan-2-yldisulfanyl)pentane
SMILES CC(C)(C)CC(C)(C)SSC(C)(C)CC(C)(C)C

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 290.57
Percent Purity ≥75.0% (GC)
Physical Form Liquid
Chemical Name or Material Di-tert-octyl Disulfide
EINECSNumber (2)-0473&(2)-0475
TSCA No