missing translation for 'onlineSavingsMsg'
Learn More

Di-t-butylmethylphosphonium tetrafluoroborate

Catalog No. AC386960050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC386960050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC386960050 Supplier Thermo Scientific Chemicals Supplier No. 386960050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 479094-62-7 | C9H22BF4P | 248.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 479094-62-7
Molecular Formula C9H22BF4P
Molecular Weight (g/mol) 248.05
MDL Number MFCD03840579
InChI Key BRDLRXCAHKUWJS-UHFFFAOYSA-O
Synonym di-tert-butyl methyl phosphonium tetrafluoroborate, di-tert-butylmethylphosphonium tetrafluoroborate, di-tert-butyl methyl phosphanium tetrafluoroborate, t-bu 2pmehbf4, di-t-butylmethylphosphonium tetrafluoroborate, bis 1,1-dimethylethyl methylphosphine tetrafluoroborate, methyl bis 2-methyl-2-propanyl phosphonium tetrafluoroborate
PubChem CID 11413958
IUPAC Name di-tert-butyl(methyl)phosphanium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.C[PH+](C(C)(C)C)C(C)(C)C

Specifications

Color White
Elemental Analysis 98.5% min.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Formula Weight 248.05
Physical Form Crystalline Powder
Chemical Name or Material Di-tert-butylmethylphosphonium tetrafluoroborate
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only