accessibility menu, dialog, popup

UserName

BFFFFH3CCH3H3CPHCH3CH3CH3H3C

CAS RN 479094-62-7

IUPAC Name: di-tert-butyl(methyl)phosphanium; tetrafluoroboranuide
Synonyms: di-tert-butyl(methyl)phosphanium tetrafluoroborate
Molecular Weight (g/mol): 248.05
Molecular Formula: C9H22BF4P
InChi Key: BRDLRXCAHKUWJS-UHFFFAOYSA-O
SMILES: F[B-](F)(F)F.C[PH+](C(C)(C)C)C(C)(C)C
Physical Form 
  • (3)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)