missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butylchlorophosphine 95.0+%, TCI America™

Catalog No. p-7127834
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36325G 5 g
D363225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36325G Supplier TCI America Supplier No. D36325G
Only null left

Chemical Identifiers

CAS 13716-10-4
Molecular Formula C8H18ClP
Molecular Weight (g/mol) 180.656
MDL Number MFCD00008815
InChI Key MCRSZLVSRGTMIH-UHFFFAOYSA-N
Synonym di-tert-butylchlorophosphine, di-tert-butylchlorophosphane, di-t-butylchlorophosphine, phosphinous chloride, bis 1,1-dimethylethyl, chlorodi-tert-butylphosphine, phosphinous chloride, di-tert-butyl, di-tert-butyl chloro phosphane, di-tert-butylphosphinous chloride, clptbu2, pubchem6477
PubChem CID 139566
IUPAC Name ditert-butyl(chloro)phosphane
SMILES CC(C)(C)P(C(C)(C)C)Cl

Specifications

Melting Point 3°C
Color Yellow
Boiling Point 72°C
Quantity 5 g
UN Number 3265
Formula Weight 180.66
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Di-tert-butylchlorophosphine