accessibility menu, dialog, popup

UserName

CAS RN 13716-10-4

IUPAC Name: di-tert-butyl(chloro)phosphane
Synonyms: di-tert-butyl(chloro)phosphane
Molecular Weight (g/mol): 180.66
Molecular Formula: C8H18ClP
InChi Key: MCRSZLVSRGTMIH-UHFFFAOYSA-N
SMILES: CC(C)(C)P(Cl)C(C)(C)C
Boiling Point 
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)