missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butylchlorophosphine, 96%

Catalog No. p-3778171
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC320900050 5 mL Glass bottle
AC320900250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC320900050 Supplier Thermo Scientific Chemicals Supplier No. 320900050
Only null left

CAS: 13716-10-4 | C8H18ClP | 180.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13716-10-4
Molecular Formula C8H18ClP
Molecular Weight (g/mol) 180.66
MDL Number MFCD00008815
InChI Key MCRSZLVSRGTMIH-UHFFFAOYSA-N
Synonym di-tert-butylchlorophosphine, di-tert-butylchlorophosphane, di-t-butylchlorophosphine, phosphinous chloride, bis 1,1-dimethylethyl, chlorodi-tert-butylphosphine, phosphinous chloride, di-tert-butyl, di-tert-butyl chloro phosphane, di-tert-butylphosphinous chloride, clptbu2, pubchem6477
PubChem CID 139566
IUPAC Name ditert-butyl(chloro)phosphane
SMILES CC(C)(C)P(C(C)(C)C)Cl

Specifications

Melting Point 2.0°C to 3.0°C
Color Colorless
Density 0.9500g/mL
Boiling Point 69.0°C to 70.0°C (10.0 mmHg)
Flash Point 61°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula [(CH3)3C]2PCl
Quantity 5 mL
Specific Gravity 0.95
Formula Weight 180.66
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Di-tert-butylchlorophosphine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

RUO – Research Use Only