missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-tert-butylchlorophosphine, 96%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 2.0°C to 3.0°C |
| Color | Colorless |
| Density | 0.9500g/mL |
| Boiling Point | 69.0°C to 70.0°C (10.0 mmHg) |
| Flash Point | 61°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 95% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | [(CH3)3C]2PCl |
| Quantity | 25 mL |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. Contact with water liberates toxic gas. |
| Precautionary Statement | GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective |
RUO – Research Use Only