Learn More
Di-tert-butyl N,N-diisopropylphosphoramidite, 95%, Thermo Scientific™
Chemical Identifiers
| CAS | 137348-86-8 |
|---|---|
| Molecular Formula | C14H32NO2P |
| Molecular Weight (g/mol) | 277.39 |
| MDL Number | MFCD00153506 |
| InChI Key | YGFLCNPXEPDANQ-UHFFFAOYSA-N |
| Synonym | di-tert-butyl diisopropylphosphoramidite, di-tert-butyl n,n-diisopropylphosphoramidite, di-t-butyl n,n-diisopropylphosphoramidite, di-tert-butoxy diisopropylamino phosphine, n-di-tert-butoxyphosphanyl-n-isopropyl-propan-2-amine, phosphoramidous acid, bis 1-methylethyl-, bis 1,1-dimethylethyl ester, bis tert-butoxy phosphino bis methylethyl amine, bis tert-butoxy phosphanyl diisopropylamine, acmc-1by13, di-tertbutyl diisopropyl-phosphoramidite |
| PubChem CID | 853005 |
| IUPAC Name | N-[bis[(2-methylpropan-2-yl)oxy]phosphanyl]-N-propan-2-ylpropan-2-amine |
| SMILES | CC(C)N(C(C)C)P(OC(C)(C)C)OC(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC426890010
|
N/A | 1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC426890050
|
N/A | 5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 137348-86-8 | |
| 277.39 | |
| YGFLCNPXEPDANQ-UHFFFAOYSA-N | |
| 853005 | |
| CC(C)N(C(C)C)P(OC(C)(C)C)OC(C)(C)C |
| C14H32NO2P | |
| MFCD00153506 | |
| di-tert-butyl diisopropylphosphoramidite, di-tert-butyl n,n-diisopropylphosphoramidite, di-t-butyl n,n-diisopropylphosphoramidite, di-tert-butoxy diisopropylamino phosphine, n-di-tert-butoxyphosphanyl-n-isopropyl-propan-2-amine, phosphoramidous acid, bis 1-methylethyl-, bis 1,1-dimethylethyl ester, bis tert-butoxy phosphino bis methylethyl amine, bis tert-butoxy phosphanyl diisopropylamine, acmc-1by13, di-tertbutyl diisopropyl-phosphoramidite | |
| N-[bis[(2-methylpropan-2-yl)oxy]phosphanyl]-N-propan-2-ylpropan-2-amine |
Specifications
| 137348-86-8 | |
| 85.0°C to 90.0°C (0.2mmHg) | |
| Authentic | |
| C14H32NO2P | |
| MFCD00153506 | |
| di-tert-butyl diisopropylphosphoramidite, di-tert-butyl n,n-diisopropylphosphoramidite, di-t-butyl n,n-diisopropylphosphoramidite, di-tert-butoxy diisopropylamino phosphine, n-di-tert-butoxyphosphanyl-n-isopropyl-propan-2-amine, phosphoramidous acid, bis 1-methylethyl-, bis 1,1-dimethylethyl ester, bis tert-butoxy phosphino bis methylethyl amine, bis tert-butoxy phosphanyl diisopropylamine, acmc-1by13, di-tertbutyl diisopropyl-phosphoramidite | |
| CC(C)N(C(C)C)P(OC(C)(C)C)OC(C)(C)C | |
| 277.39 | |
| 277.39 | |
| Di-tert-butyl N, N-diisopropylphosphoramidite |
| 0.8790g/mL | |
| 113°C | |
| 94% min. (GC) | |
| 1.4390 to 1.4410 (20°C, 589nm) | |
| 0.879 | |
| YGFLCNPXEPDANQ-UHFFFAOYSA-N | |
| N-[bis[(2-methylpropan-2-yl)oxy]phosphanyl]-N-propan-2-ylpropan-2-amine | |
| 853005 | |
| 95% |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
RUO – Research Use Only