missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl N,N-diisopropylphosphoramidite, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 426890050

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC426890050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

137348-86-8
277.39
YGFLCNPXEPDANQ-UHFFFAOYSA-N
853005
CC(C)N(C(C)C)P(OC(C)(C)C)OC(C)(C)C
C14H32NO2P
MFCD00153506
di-tert-butyl diisopropylphosphoramidite, di-tert-butyl n,n-diisopropylphosphoramidite, di-t-butyl n,n-diisopropylphosphoramidite, di-tert-butoxy diisopropylamino phosphine, n-di-tert-butoxyphosphanyl-n-isopropyl-propan-2-amine, phosphoramidous acid, bis 1-methylethyl-, bis 1,1-dimethylethyl ester, bis tert-butoxy phosphino bis methylethyl amine, bis tert-butoxy phosphanyl diisopropylamine, acmc-1by13, di-tertbutyl diisopropyl-phosphoramidite
N-[bis[(2-methylpropan-2-yl)oxy]phosphanyl]-N-propan-2-ylpropan-2-amine

Specifications

Di-tert-butyl N, N-diisopropylphosphoramidite
0.8790g/mL
113°C
94% min. (GC)
C14H32NO2P
5g
0.879
YGFLCNPXEPDANQ-UHFFFAOYSA-N
N-[bis[(2-methylpropan-2-yl)oxy]phosphanyl]-N-propan-2-ylpropan-2-amine
853005
95%
137348-86-8
85.0°C to 90.0°C (0.2mmHg)
Authentic
Glass bottle
1.4390 to 1.4410 (20°C, 589nm)
MFCD00153506
di-tert-butyl diisopropylphosphoramidite, di-tert-butyl n,n-diisopropylphosphoramidite, di-t-butyl n,n-diisopropylphosphoramidite, di-tert-butoxy diisopropylamino phosphine, n-di-tert-butoxyphosphanyl-n-isopropyl-propan-2-amine, phosphoramidous acid, bis 1-methylethyl-, bis 1,1-dimethylethyl ester, bis tert-butoxy phosphino bis methylethyl amine, bis tert-butoxy phosphanyl diisopropylamine, acmc-1by13, di-tertbutyl diisopropyl-phosphoramidite
CC(C)N(C(C)C)P(OC(C)(C)C)OC(C)(C)C
277.39
277.39
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions