missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl(methyl)phosphonium Tetrafluoroborate 98.0+%, TCI America™

Catalog No. D47315G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47315G 5 g
D47311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47315G Supplier TCI America Supplier No. D47315G

Chemical Identifiers

CAS 479094-62-7
Molecular Formula C9H22BF4P
Molecular Weight (g/mol) 248.05
MDL Number MFCD03840579
InChI Key BRDLRXCAHKUWJS-UHFFFAOYSA-O
Synonym di-tert-butyl methyl phosphonium tetrafluoroborate, di-tert-butylmethylphosphonium tetrafluoroborate, di-tert-butyl methyl phosphanium tetrafluoroborate, t-bu 2pmehbf4, di-t-butylmethylphosphonium tetrafluoroborate, bis 1,1-dimethylethyl methylphosphine tetrafluoroborate, methyl bis 2-methyl-2-propanyl phosphonium tetrafluoroborate
PubChem CID 11413958
IUPAC Name di-tert-butyl(methyl)phosphanium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.C[PH+](C(C)(C)C)C(C)(C)C

Specifications

Color White
Quantity 5 g
UN Number 1759
Formula Weight 248.05
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Di-tert-butyl(methyl)phosphonium Tetrafluoroborate