missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-tert-butyl Iminodicarboxylate 95.0+%, TCI America™
Supplier: TCI America I049725G
| Quantity | 25 g |
|---|
Chemical Identifiers
| 51779-32-9 | |
| 217.27 | |
| XCAQIUOFDMREBA-UHFFFAOYSA-N | |
| 279800 | |
| CC(C)(C)OC(=O)NC(=O)OC(C)(C)C |
| C10H19NO4 | |
| MFCD00043309 | |
| di-tert-butyl iminodicarboxylate, di-t-butyl iminodicarboxylate, di-tert-butyl imidodicarbonate, n-boc-tert-butylcarbamate, tert-butyl iminodicarboxylate, di-tert-butyl iminodicarbonate, di-tert-butyl-iminodicarboxylate, iminodicarboxylic acid di-tert-butyl ester, tert-butyl n-tert-butoxy carbonyl carbamate, di tert-butyl imidodicarbonate | |
| tert-butyl N-[(tert-butoxy)carbonyl]carbamate |
Specifications
| 117°C | |
| 51779-32-9 | |
| C10H19NO4 | |
| di-tert-butyl iminodicarboxylate, di-t-butyl iminodicarboxylate, di-tert-butyl imidodicarbonate, n-boc-tert-butylcarbamate, tert-butyl iminodicarboxylate, di-tert-butyl iminodicarbonate, di-tert-butyl-iminodicarboxylate, iminodicarboxylic acid di-tert-butyl ester, tert-butyl n-tert-butoxy carbonyl carbamate, di tert-butyl imidodicarbonate | |
| CC(C)(C)OC(=O)NC(=O)OC(C)(C)C | |
| 25 g | |
| 279800 | |
| ≥95.0% (GC) |
| Di-tert-butyl Iminodicarboxylate | |
| White-Yellow | |
| MFCD00043309 | |
| XCAQIUOFDMREBA-UHFFFAOYSA-N | |
| tert-butyl N-[(tert-butoxy)carbonyl]carbamate | |
| 217.27 | |
| 217.27 | |
| Crystalline Powder |
Safety and Handling
TSCA : No