missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl Iminodicarboxylate 95.0+%, TCI America™

Catalog No. p-7131442
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04975G 5 g
I049725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I04975G Supplier TCI America Supplier No. I04975G

Chemical Identifiers

CAS 51779-32-9
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.27
MDL Number MFCD00043309
InChI Key XCAQIUOFDMREBA-UHFFFAOYSA-N
Synonym di-tert-butyl iminodicarboxylate, di-t-butyl iminodicarboxylate, di-tert-butyl imidodicarbonate, n-boc-tert-butylcarbamate, tert-butyl iminodicarboxylate, di-tert-butyl iminodicarbonate, di-tert-butyl-iminodicarboxylate, iminodicarboxylic acid di-tert-butyl ester, tert-butyl n-tert-butoxy carbonyl carbamate, di tert-butyl imidodicarbonate
PubChem CID 279800
IUPAC Name tert-butyl N-[(tert-butoxy)carbonyl]carbamate
SMILES CC(C)(C)OC(=O)NC(=O)OC(C)(C)C

Specifications

Melting Point 117°C
Color White-Yellow
Quantity 5 g
Formula Weight 217.27
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Di-tert-butyl Iminodicarboxylate