missing translation for 'onlineSavingsMsg'
Learn More

[Di-tert-butyl(chloro)phosphine]palladium(II) Dichloride Dimer 97.0+%, TCI America™

Catalog No. D3704100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3704100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D3704100MG Supplier TCI America Supplier No. D3704100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 386706-33-8
Molecular Formula C16H36Cl6P2Pd2
Molecular Weight (g/mol) 715.952
MDL Number MFCD05664440
InChI Key NJIMMSLZDJFGSV-UHFFFAOYSA-J
Synonym Dichloro[di-tert-butyl(chloro)phosphine]palladium(II) Dimer
PubChem CID 53384540
IUPAC Name ditert-butyl(chloro)phosphane;palladium(2+);tetrachloride
SMILES CC(C)(C)P(C(C)(C)C)Cl.CC(C)(C)P(C(C)(C)C)Cl.[Cl-].[Cl-].[Cl-].[Cl-].[Pd+2].[Pd+2]

Specifications

Melting Point 222°C
Quantity 100 mg
UN Number 3261
Formula Weight 715.95
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material [Di-tert-butyl(chloro)phosphine]palladium(II) Dichloride Dimer