missing translation for 'onlineSavingsMsg'
Learn More

Di-n-propyl sulfide, 98+%

CAS: 111-47-7 | C6H14S | 118.24 g/mol

$148.81 - $1609.96

Chemical Identifiers

CAS 111-47-7
Molecular Formula C6H14S
Molecular Weight (g/mol) 118.24
MDL Number MFCD00009379
InChI Key ZERULLAPCVRMCO-UHFFFAOYSA-N
Synonym propyl sulfide, dipropyl sulfide, di-n-propyl sulfide, propane, 1,1'-thiobis, dipropyl thioether, propyl monosulfide, dipropyl sulphide, n-propyl sulfide, 4-thiaheptane, 1,1'-thiobispropane
PubChem CID 8118
IUPAC Name 1-propylsulfanylpropane
SMILES CCCSCCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2315514
View Documents
Thermo Scientific Chemicals
B2315514
25 g
Each for $148.81
Only null left
 
AAB2315522
View Documents
Thermo Scientific Chemicals
B2315522
100 g
Each for $416.62
Only null left
 
AAB2315536
View Documents
Thermo Scientific Chemicals
B2315536
500 g
Each for $1,609.96
Only null left
 
Description

Description

Di-n-propyl sulfide is used in organic synthesis and used as a daily fragrance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-propyl sulfide is used in organic synthesis and used as a daily fragrance.

Solubility
Soluble in water 351mg/L @ 25°C.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Specifications

Chemical Identifiers

111-47-7
118.24
ZERULLAPCVRMCO-UHFFFAOYSA-N
8118
CCCSCCC
C6H14S
MFCD00009379
propyl sulfide, dipropyl sulfide, di-n-propyl sulfide, propane, 1,1'-thiobis, dipropyl thioether, propyl monosulfide, dipropyl sulphide, n-propyl sulfide, 4-thiaheptane, 1,1'-thiobispropane
1-propylsulfanylpropane

Specifications

111-47-7
0.838
28°C (82°F)
C6H14S
MFCD00009379
UN1993
14,7868
Soluble in water 351mg/L @ 25°C.
CCCSCCC
118.24
118.24
Di-n-propyl sulfide
-102°C
141°C to 142°C
Stench
1.4487
25 g
1719002
propyl sulfide, dipropyl sulfide, di-n-propyl sulfide, propane, 1,1'-thiobis, dipropyl thioether, propyl monosulfide, dipropyl sulphide, n-propyl sulfide, 4-thiaheptane, 1,1'-thiobispropane
ZERULLAPCVRMCO-UHFFFAOYSA-N
1-propylsulfanylpropane
8118
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 203-873-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only