missing translation for 'onlineSavingsMsg'
Learn More

Di-n-propyl sulfide, 98+%

Catalog No. AAB2315514
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2315514 25 g
AAB2315522 100 g
AAB2315536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2315514 Supplier Thermo Scientific Chemicals Supplier No. B2315514

CAS: 111-47-7 | C6H14S | 118.24 g/mol

Di-n-propyl sulfide is used in organic synthesis and used as a daily fragrance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-propyl sulfide is used in organic synthesis and used as a daily fragrance.

Solubility
Soluble in water 351mg/L @ 25°C.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 111-47-7
Molecular Formula C6H14S
Molecular Weight (g/mol) 118.24
MDL Number MFCD00009379
InChI Key ZERULLAPCVRMCO-UHFFFAOYSA-N
Synonym propyl sulfide, dipropyl sulfide, di-n-propyl sulfide, propane, 1,1'-thiobis, dipropyl thioether, propyl monosulfide, dipropyl sulphide, n-propyl sulfide, 4-thiaheptane, 1,1'-thiobispropane
PubChem CID 8118
IUPAC Name 1-propylsulfanylpropane
SMILES CCCSCCC

Specifications

Melting Point -102°C
Density 0.838
Boiling Point 141°C to 142°C
Flash Point 28°C (82°F)
Odor Stench
Refractive Index 1.4487
Quantity 25 g
UN Number UN1993
Beilstein 1719002
Merck Index 14,7868
Solubility Information Soluble in water 351mg/L @ 25°C.
Formula Weight 118.24
Percent Purity ≥98%
Chemical Name or Material Di-n-propyl sulfide
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 203-873-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only