missing translation for 'onlineSavingsMsg'
Learn More

Di-n-propyl phthalate, 98%

Catalog No. p-7027360
Change view
Click to view available options
Quantity:
250 g
50 g
Catalog No. Quantity
AAB2405618 50 g
AAB2405630 250 g
2 options

Catalog No. AAB2405618

Supplier: Thermo Scientific Chemicals B2405618

Only null left

CAS: 131-16-8 | C14H18O4 | 250.294 g/mol

Di-n-propyl phthalate is used to make plasticizers and polymer additives. It is also used in chemical reagents, organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-propyl phthalate is used to make plasticizers and polymer additives. It is also used in chemical reagents, organic intermediates.

Solubility
Soluble in chloroform, methanol.

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 131-16-8
Molecular Formula C14H18O4
Molecular Weight (g/mol) 250.294
MDL Number MFCD00009371
InChI Key MQHNKCZKNAJROC-UHFFFAOYSA-N
Synonym dipropyl phthalate, di-n-propyl phthalate, di-n-propylphthalate, phthalic acid dipropyl ester, phthalic acid, dipropyl ester, phthalic acid di-n-propyl ester, 1,2-benzenedicarboxylic acid, dipropyl ester, dipropylphthalate, n-dipropyl phthalate, unii-8usp5y77ss
PubChem CID 8559
ChEBI CHEBI:60069
IUPAC Name dipropyl benzene-1,2-dicarboxylate
SMILES CCCOC(=O)C1=CC=CC=C1C(=O)OCCC

Specifications

Density 1.082
Boiling Point 168°C to 170°C (3 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.497
Quantity 50 g
UN Number UN3082
Beilstein 2332522
Solubility Information Soluble in chloroform,methanol.
Formula Weight 250.3
Percent Purity 98%
Chemical Name or Material Di-n-propyl phthalate
Show More Show Less
Hazard Category H351-H361
Hazard Statement GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P280-P308+P313-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 205-015-8
RTECSNumber TI1940000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only