missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyldichlorosilane, 97%

Catalog No. p-7052124
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4259909 10 g
AA4259918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4259909 Supplier Thermo Scientific Chemicals Supplier No. 04259909

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18416-07-4 | C16H34Cl2Si | 325.433 g/mol

Di-n-octyldichlorosilane is used in the synthesis of organic photovoltaics. It is also used in medicine field, electronic industry and polymer material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18416-07-4
Molecular Formula C16H34Cl2Si
Molecular Weight (g/mol) 325.433
MDL Number MFCD01074472
InChI Key CVAGYCLEIYGJQT-UHFFFAOYSA-N
Synonym di-n-octyldichlorosilane, dichloro dioctyl silane, silane, dichlorodioctyl, dioctyldichlorosilane, dioctyldichlorsilan, dichlorodi-n-octylsilane, di-n-octyldichlorosilane 10g
PubChem CID 248429
IUPAC Name dichloro(dioctyl)silane
SMILES CCCCCCCC[Si](CCCCCCCC)(Cl)Cl

Specifications

Boiling Point 145°C (0.2mmHg)
Color Colorless to Yellow
Physical Form Liquid
Quantity 10 g
Linear Formula [CH3(CH2)7]2SiCl2
UN Number UN2987
Solubility Information Sparingly Soluble (8.8E-6g/L) at 25°C.
Formula Weight 325.44
Percent Purity 97%
Sensitivity Moisture sensitive
Density 0.940 g/mL
Chemical Name or Material Di-n-octyldichlorosilane
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only