missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyl Sulfoxide 98.0+%, TCI America™

Catalog No. D29651G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29651G 1 g
D29655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29651G Supplier TCI America Supplier No. D29651G

Chemical Identifiers

CAS 1986-89-6
Molecular Formula C16H34OS
Molecular Weight (g/mol) 274.507
MDL Number MFCD00039514
InChI Key VWCFQNQVNVMFGV-UHFFFAOYSA-N
Synonym n-Octyl Sulfoxide
PubChem CID 74805
IUPAC Name 1-octylsulfinyloctane
SMILES CCCCCCCCS(=O)CCCCCCCC

Specifications

Melting Point 71°C
Color White
Quantity 1 g
Formula Weight 274.51
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Di-n-octyl Sulfoxide