missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyl sulfide, 97%

Catalog No. AAA1795536
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1795536 500 g
AAA1795514 25 g
AAA1795522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1795536 Supplier Thermo Scientific Chemicals Supplier No. A1795536
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2690-08-6 | C16H34S | 258.51 g/mol

It is employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a pharmaceutical intermediate.

Solubility
Solubility in water, 0.004678 mg/L @ 25°C (est)

Notes
Air & Moisture Sensitive. Store away from acids, oxidizing agents, air, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 2690-08-6
MDL Number MFCD00009566

Specifications

Melting Point 0°C to 1°C
Density 0.844
Boiling Point 179°C to 180°C (10 mmHg)
Flash Point 160°C (320°F)
Odor Stench
Molecular Formula C12H17NO
Refractive Index 1.4624
Quantity 500 g
Beilstein 1701624
Synonym benzamide, n,n-diethyl-4-methyl, p-deta, n,n-diethyl-p-toluamide, p-toluamide, n,n-diethyl, p-deet, unii-i8bwy8k43u, i8bwy8k43u, n,n-diethyl-4-methyl-benzamide, p-toluamide,n-diethyl, n,n-diethyl-4-toluamide
InChI Key PUZORFQMRDHKBT-UHFFFAOYSA-N
SMILES CCN(CC)C(=O)C1=CC=C(C=C1)C
IUPAC Name N,N-diethyl-4-methylbenzamide
Molecular Weight (g/mol) 191.27
PubChem CID 75946
Formula Weight 258.51
Percent Purity 97%
Chemical Name or Material Di-n-octyl sulfide
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-259-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only